This product is now discontinued, View Related Products
A300-Series Multi-Channel Pipettors
Fully autoclavable with enhanced UV resistance
  • Ergonomic, comfortable, and light weight
  • Superior accuracy and precision
  • Locking counter prevents accidental volume fluctuations
  • Easily identify your pipette using detachable color-coded drops
  • Reverse pipetting option ensures accurate, precise aspirating and dispensing of viscous liquids
  • Calibration stability guarantees accuracy of dispense volume even after multiple autoclaving and pipetting
  • Enhanced UV resistance provides added protection from biohazardous elements
  • Universal tip compatibility to use with wide range of tips up to 1000μL
  • Meets DIN EN ISO 8655 standard
Cole-Parmer A300-series Multi-Channel Pipettors have been ergonomically designed to the most rigorous standards for accuracy, precision and comfort. Pipettor’s user-friendly handle design with soft grip gives ease of operation and reduces fatigue. Its’ light weight and low plunger force minimizes RSI (Repetitive Strain Injury). Sturdy PVDF tip cone ensures a high resistance against chemicals. Blow out system ensures last drop dispensing thus preventing critical sample loss. Innovative soft grip protects heat transfer to internal components ensuring accurate operation.
24,300.00 - ₹33,500.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Volume (µL) Min Volume (µL) Number Of Channels
Availability Pricing
COMPARE
10 0.5 8
₹24,300.00INR / Each
COMPARE
50 5 8
₹24,300.00INR / Each
COMPARE
300 30 8
₹24,300.00INR / Each
COMPARE
10 0.5 12
₹33,500.00INR / Each
COMPARE
50 5 12
₹33,500.00INR / Each
COMPARE
300 30 12
₹33,500.00INR / Each