This product is now discontinued, View Related Products
A200-Series Single Channel Fixed-Volume Pipettors
Fully autoclavable with enhanced UV resistance
  • Ergonomic, comfortable, and light weight
  • Superior accuracy and precision
  • Locking counter prevents accidental volume fluctuations
  • Easily identify your pipette using detachable color-coded drops
  • Reverse pipetting option ensures accurate, precise aspirating and dispensing of viscous liquids
  • Calibration stability guarantees accuracy of dispense volume even after multiple autoclaving and pipetting
  • Enhanced UV resistance provides added protection from biohazardous elements
  • Universal tip compatibility to use with wide range of tips up to 1000μL
  • Meets DIN EN ISO 8655 standard
Cole-Parmer A200-series Single Channel Fixed-Volume Pipettors have been ergonomically designed to the most rigorous standards for accuracy, precision and comfort. Pipettor’s user-friendly handle design with soft grip gives ease of operation and reduces fatigue. Its’ light weight and low plunger force minimizes RSI (Repetitive Strain Injury). Sturdy PVDF tip cone ensures a high resistance against chemicals. Blow out system ensures last drop dispensing thus preventing critical sample loss. Innovative soft grip protects heat transfer to internal components ensuring accurate operation.
2,700.00 - ₹9,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Accuracy (%) Max Volume (µL) Number Of Channels
Availability Pricing
COMPARE
±1.3 5 1
₹6,000.00INR / Each
COMPARE
±0.5 50 1
₹6,000.00INR / Each
COMPARE
±0.5 100 1
₹6,000.00INR / Each
COMPARE
±0.3 500 1
₹2,700.00INR / Each
COMPARE
±0.3 1000 1
₹9,000.00INR / Each
COMPARE
±0.3 5000 1
₹2,700.00INR / Each
COMPARE
±0.6 10000 1
₹5,000.00INR / Each
Showing 1- 7 of 7