This product is now discontinued, View Related Products
A100-Series Single Channel Adjustable-Volume Pipettors
Fully autoclavable with enhanced UV resistance
  • Ergonomic, comfortable, and light weight
  • Superior accuracy and precision
  • Locking counter prevents accidental volume fluctuations
  • Easily identify your pipette using detachable color-coded drops
  • Reverse pipetting option ensures accurate, precise aspirating and dispensing of viscous liquids
  • Calibration stability guarantees accuracy of dispense volume even after multiple autoclaving and pipetting
  • Enhanced UV resistance provides added protection from biohazardous elements
  • Universal tip compatibility to use with wide range of tips up to 1000μL
  • Meets DIN EN ISO 8655 standard
Cole-Parmer A100-series Single Channel Adjustable-Volume Pipettors have been ergonomically designed to the most rigorous standards for accuracy, precision and comfort. Pipettor’s user-friendly handle design with soft grip gives ease of operation and reduces fatigue. Its’ light weight and low plunger force minimizes RSI (Repetitive Strain Injury). Sturdy PVDF tip cone ensures a high resistance against chemicals. Blow out system ensures last drop dispensing thus preventing critical sample loss. Innovative soft grip protects heat transfer to internal components ensuring accurate operation.
4,600.00 - ₹9,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Volume (µL) Min Volume (µL) Number Of Channels
Availability Pricing
COMPARE
2.5 0.1 1
₹8,000.00INR / Each
COMPARE
10 0.5 1
₹9,000.00INR / Each
COMPARE
50 5 1
₹8,000.00INR / Each
COMPARE
100 10 1
₹7,000.00INR / Each
COMPARE
200 20 1
₹7,000.00INR / Each
COMPARE
1000 100 1
₹9,000.00INR / Each
COMPARE
5000 500 1
₹9,000.00INR / Each
COMPARE
10000 2000 1
₹4,600.00INR / Each
Showing 1- 8 of 8