This product is now discontinued, View Related Products
A&D GX Toploading Balances
Super Hybrid Sensor (SHS) technology offers fastest response time in its class – one second!
  • Never lose data again — store weighing data in non-volatile memory
  • Complementary WinCT software supports data collection and statistical analysis
  • Save power with the auto power-off function after 10 minutes of inactivity
  • Internal calibration saves time and money with quick calibration
  • Automatically self-calibrates when ambient temperature changes are detected
  • Calibrate on-demand with one-touch automatic calibration
These user-friendly balances feature Super Hybrid Sensor (SHS) technology which improves response speed, accuracy, and minimizes costs. SHS combines the best of magnetic force restoration and single point parallelogram load sensing technologies. Its unique design provides the fastest response speed in its class – just one second.
The GX-series toploading balances feature USB and RS-232 interface for transfer of weighing data to a computer or optional printer. The complementary software is easy-to-use and is designed to download information from your A&D balance directly into your personal computer, saving you the time and effort of having to manually enter your weighing data and reducing the chance of data input errors. Compliant with GLP, GMP, GCP, and ISO output, these balances will output the manufacturer, model, serial number, ID number, space for signature, calibration records, and other pertinent information, for documentation requirements.
435,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
A&D Weighing GX-600 Precision Toploading Balance, Internal Calibration; 610 g x 0.001 g WW-11110-57 Mfr # GX-600
COMPARE
610 115 0.001
Call For Price
A&D Weighing GX-800 Precision Toploading Balance, Internal Calibration; 810 g x 0.001 g WW-11110-58 Mfr # GX-800
COMPARE
810 115 / 230 0.001
Call For Price
A&D Weighing GX-1000 Precision Toploading Balance, Internal Calibration; 1100 g x 0.001 g WW-11110-59 Mfr # GX-1000
COMPARE
1100 115 / 230 0.001
Call For Price
A&D Weighing GX-200 Precision Toploading Balance, Internal Calibration; 210 g x 0.001 g WW-11118-00 Mfr # GX-200
COMPARE
210 115 / 230 0.001
Call For Price
A&D Weighing GX-400 Precision Toploading Balance, Internal Calibration; 410 g x 0.001 g WW-11118-02 Mfr # GX-400
COMPARE
410 115 / 230 0.001
₹435,000.00INR / Each
A&D Weighing GX-600 Toploading Balance, 610g x 0.01g Internal Calibration WW-11118-04 Mfr # GX-600
COMPARE
610 115 0.01
Call For Price
A&D Weighing GX-2000 Precision Toploading Balance, Internal Calibration; 2100 g x 0.01 g WW-11118-06 Mfr # GX-2000
COMPARE
2100 115 / 230 0.01
Call For Price
A&D Weighing GX-4000 Precision Toploading Balance, Internal Calibration; 4100 g x 0.01 g WW-11118-08 Mfr # GX-4000
COMPARE
4100 115 / 230 0.01
Call For Price
A&D Weighing GX-6000 Precision Toploading Balance, Internal Calibration; 6100 g x 0.1 g WW-11118-10 Mfr # GX-6000
COMPARE
6100 115 / 230 0.1
Call For Price
A&D Weighing GX-6100 Precision Toploading Balance, Internal Calibration; 6100 g x 0.01 g WW-11118-12 Mfr # GX-6100
COMPARE
6100 115 / 230 0.01
Call For Price
A&D Weighing GX-8000 Precision Toploading Balance, Internal Calibration; 8100 g x 0.1 g WW-11118-14 Mfr # GX-8000
COMPARE
8100 115 / 230 0.1
Call For Price
Showing 1- 11 of 11