This product is now discontinued, View Related Products
A&D GF Toploading Balances
Super Hybrid Sensor (SHS) technology offers fastest response time in its class – one second!
  • Never lose data again — store weighing data in non-volatile memory
  • Complementary WinCT software supports data collection and statistical analysis
  • Save power with the auto power-off function after 10 minutes of inactivity
  • External calibration
These user-friendly balances feature Super Hybrid Sensor (SHS) technology which improves response speed, accuracy, and minimizes costs. SHS combines the best of magnetic force restoration and single point parallelogram load sensing technologies. Its unique design provides the fastest response speed in its class – just one second.
The GF-series toploading balances feature RS-232 interface for transfer of weighing data to a computer or optional printer. The complementary software is easy-to-use and is designed to download information from your A&D balance directly into your personal computer, saving you the time and effort of having to manually enter your weighing data and reducing the chance of data input errors. Compliant with GLP, GMP, GCP, and ISO output, these balances will output the manufacturer, model, serial number, ID number, space for signature, calibration records, and other pertinent information, for documentation requirements.
516,000.00INR / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
A&D Weighing GF-400 Precision Toploading Balance, External Calibration; 410 g x 0.001 g WW-11110-61 Mfr # GF-400
COMPARE
410 115 / 230 0.001
Call For Price
A&D Weighing GF-600 Precision Toploading Balance, External Calibration; 610 g x 0.001 g WW-11110-62 Mfr # GF-600
COMPARE
610 115 / 230 0.001
Call For Price
A&D Weighing GF-800 Precision Toploading Balance, External Calibration; 810 g x 0.001 g WW-11110-63 Mfr # GF-800
COMPARE
810 115 / 230 0.001
Call For Price
A&D Weighing GF-1000 Precision Toploading Balance, External Calibration; 1100 g x 0.001 g WW-11110-64 Mfr # GF-1000
COMPARE
1100 115 / 230 0.001
Call For Price
A&D Weighing GF-4000 Precision Toploading Balance, External Calibration; 4100 g x 0.01 g WW-11110-65 Mfr # GF-4000
COMPARE
4100 115 / 230 0.01
Call For Price
A&D Weighing GF-6100 Precision Toploading Balance, External Calibration; 6100 g x 0.01 g WW-11110-66 Mfr # GF-6100
COMPARE
6100 115 / 230 0.01
Call For Price
A&D Weighing GF-200 Precision Toploading Balance, External Calibration; 210 g x 0.001 g WW-11133-00 Mfr # GF-200
COMPARE
210 115 / 230 0.001
Call For Price
A&D Weighing GF-300 Precision Toploading Balance, External Calibration; 310 g x 0.001 g WW-11133-02 Mfr # GF-300
COMPARE
310 115 / 230 0.001
Call For Price
A&D Weighing GF-1200 Precision Toploading Balance, External Calibration; 1210 g x 0.01 g WW-11133-06 Mfr # GF-1200
COMPARE
1210 115 / 230 0.01
Call For Price
A&D Weighing GF-2000 Precision Toploading Balance, External Calibration; 2100 g x 0.01 g WW-11133-08 Mfr # GF-2000
COMPARE
2100 115 / 230 0.01
Call For Price
A&D Weighing GF-3000 Precision Toploading Balance, External Calibration; 3100 g x 0.01 g WW-11133-10 Mfr # GF-3000
COMPARE
3100 115 / 230 0.01
Call For Price
A&D Weighing GF-6000 Precision Toploading Balance, External Calibration; 6100 g x 0.1 g WW-11133-12 Mfr # GF-6000
COMPARE
6100 115 / 230 0.1
Call For Price
A&D Weighing GF-8K Precision High Capacity Toploading Balance, External Calibration; 8.1 kg x 0.01 g WW-11229-68 Mfr # GF-8K
COMPARE
8100 115 0.01
Call For Price
A&D Weighing GF-8K2 Precision High Capacity Toploading Balance, External Calibration; 8.1/2.1 kg x 0.1/0.01 g WW-11229-69 Mfr # GF-8K2
COMPARE
8100 115 0.1
Call For Price
A&D Weighing GF-10K Precision High Capacity Toploading Balance, External Calibration; 10.1 kg x 0.01 g WW-11229-70 Mfr # GF-10K
COMPARE
10100 115 0.01
Call For Price
A&D Weighing GF20K High Capacity Toploading Balance, 20kg x 0.1g WW-11229-72 Mfr # GF20K
COMPARE
21000 115 0.1
Call For Price
A&D Weighing GF-30K Precision High Capacity Toploading Balance, External Calibration; 31 kg x 0.1 g WW-11229-73 Mfr # GF-30K
COMPARE
31000 115 0.1
Call For Price
A&D Weighing GF-32K Precision High Capacity Toploading Balance, External Calibration; 31/6.1 kg x 1/0.1 g WW-11229-74 Mfr # GF-32K
COMPARE
31000 115 1
₹516,000.00INR / Each
Showing 1- 18 of 18