A&D Apollo GF-A Toploading Balances
Super Hybrid Sensor (SHS) technology offers fastest response time in its class – one second!
  • Streamline your dosing and filling applications with flow rate display (FRD)
  • Never lose data again — store weighing data in non-volatile memory
  • Complementary WinCT software supports data collection and statistical analysis
  • Save power with the auto power-off function after 10 minutes of inactivity
  • Easy one touch external calibration performs and documents calibration
These user-friendly balances feature Super Hybrid Sensor (SHS) technology which improves response speed, accuracy, and minimizes costs. SHS combines the best of magnetic force restoration and single point parallelogram load sensing technologies. Its unique design provides the fastest response speed in its class – just one second.
The GF Apollo series toploading balances also provide innovative features for advanced operation. Electronically controlled Load (ECL) performs a self-diagnostic test preceded by an internal repeatability test of you balance without an external weight. The standard deviation (SD) check allows for daily precision checks — ideal for SOPs. The Min-S alert ensures that that balance is operating within USP41 compliance.
Impact shock detection (ISD) visually and audibly indicates the magnitude of impacts shocks. The balance will then document the date, time, and shock level to provide real-time feedback and prevent damage of the sensor.
Flow rate display (FRD) enhances accuracy and precision of dosing and filling applications. FR-Cal calculates, displays, and outputs the mass or volume flow rate of your dispensing application via density. The comparator then provides feedback if the flow rate is within desired limits. The tare values are then stored in nonvolatile memory if an application was interrupted.
249,000.00 - ₹610,000.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pan Length (in) Capacity (g) Readability (g)
Availability Pricing
A&D Weighing GF-123A Apollo Precision Toploading Balance, External Calibration; 122 g x 0.001 g WW-11109-15 Mfr # GF-123A
COMPARE
5 122 0.001
₹249,000.00INR / Each
A&D Weighing GF-203A Apollo Precision Toploading Balance, External Calibration; 220 g x 0.001 g WW-11109-16 Mfr # GF-203A
COMPARE
5 220 0.001
₹274,000.00INR / Each
A&D Weighing GF-303A Apollo Precision Toploading Balance, External Calibration; 320 g x 0.001 g WW-11109-17 Mfr # GF-303A
COMPARE
5 320 0.001
₹309,000.00INR / Each
A&D Weighing GF-403A Apollo Precision Toploading Balance, External Calibration; 420 g x 0.001 g WW-11109-18 Mfr # GF-403A
COMPARE
5 420 0.001
₹335,000.00INR / Each
A&D Weighing GF-603A Apollo Precision Toploading Balance, External Calibration; 620 g x 0.001 g WW-11109-19 Mfr # GF-603A
COMPARE
5 620 0.001
₹395,000.00INR / Each
A&D Weighing GF-1003A Apollo Precision Toploading Balance, External Calibration; 1100 g x 0.001 g WW-11109-21 Mfr # GF-1003A
COMPARE
5 1100 0.001
₹474,000.00INR / Each
A&D Weighing GF-1603A Apollo Precision Toploading Balance, External Calibration; 1620 g x 0.001 g WW-11109-22 Mfr # GF-1603A
COMPARE
5 1620 0.001
₹610,000.00INR / Each
A&D Weighing GF-1202A Apollo Precision Toploading Balance, External Calibration; 1220 g x 0.01 g WW-11109-23 Mfr # GF-1202A
COMPARE
6 1/2 1220 0.01
₹278,000.00INR / Each
A&D Weighing GF-2002A Apollo Precision Toploading Balance, External Calibration; 2200 g x 0.01 g WW-11109-24 Mfr # GF-2002A
COMPARE
6 1/2 2200 0.01
₹285,000.00INR / Each
A&D Weighing GF-3002A Apollo Precision Toploading Balance, External Calibration; 3200 g x 0.01 g WW-11109-25 Mfr # GF-3002A
COMPARE
6 1/2 3200 0.01
₹294,000.00INR / Each
A&D Weighing GF-4002A Apollo Precision Toploading Balance, External Calibration; 4200 g x 0.01 g WW-11109-26 Mfr # GF-4002A
COMPARE
6 1/2 4200 0.01
₹378,000.00INR / Each
A&D Weighing GF-6002A Apollo Precision Toploading Balance, External Calibration; 6200 g x 0.01 g WW-11109-27 Mfr # GF-6002A
COMPARE
6 1/2 6200 0.01
₹461,000.00INR / Each
A&D Weighing GF-10002A Apollo Precision Toploading Balance, External Calibration; 10200 g x 0.01 g WW-11109-28 Mfr # GF-10002A
COMPARE
6 1/2 10200 0.01
₹569,000.00INR / Each
A&D Weighing GF-6001A Apollo Precision Toploading Balance, External Calibration; 6200 g x 0.1 g WW-11109-29 Mfr # GF-6001A
COMPARE
6 1/2 6200 0.1
₹314,000.00INR / Each
A&D Weighing GF-10001A Apollo Precision Toploading Balance, External Calibration; 10200 g x 0.1 g WW-11109-31 Mfr # GF-10001A
COMPARE
6 1/2 10200 0.1
₹329,000.00INR / Each
Showing 1- 15 of 15