Lascar Electronics 4 to 20 mA Loop Powered Panel Meters
Special installation tools are not required for current measurement
  • No soldering needed—connections are made through screw terminals
  • Snap-in and bezel mounting design makes installation quick and easy
  • Calibrates and scales rapidly with built in potentiometers
  • Displays are back litfor a clear display, even in low light conditions
Compact, loop-powered panel meters provide an accurate, zero corrected indication of 4 to 20 mA current readings. Multi-turn potentiometers allow sensitive adjustment and scaling of the indicator, and decimal point placement may be selected through a jumper link. Waterproof IP 67 or NEMA 4X rating can be achieved with the mounting bezel accessory.
21,200.00 - ₹26,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Input Type
Availability Pricing
Lascar Electronics DPM 342 4-20mA Loop Powered Panel Meter; 0.43" Digits WW-65595-00 Mfr # DPM 342
COMPARE
₹21,200.00INR / Each
Lascar Electronics DPM 742-BL 4-20mA Loop Powered Panel Meter; 0.5" Digits WW-65595-01 Mfr # DPM 742-BL
COMPARE
₹24,100.00INR / Each
Lascar Electronics DPM 942 4-20mA Loop Powered Panel Meter; 0.75" Digits WW-65595-02 Mfr # DPM 942
COMPARE
₹25,300.00INR / Each
Lascar Electronics DPM 942-FPSI 4-20mA Loop Panel Meter with Programmable Red/Green Backlighting WW-65595-03 Mfr # DPM 942-FPSI
COMPARE
₹26,400.00INR / Each