This product is now discontinued, View Related Products
3M™ Organic Vapor Monitors
Keep workers safe from hazardous environments
  • Simple and effective — badge-style design ensures easy compliance with air monitoring reulations
The 3M™ Organic Vapor Passive Air Monitoring badges use a single charcoal sorbent wafer for easy collection of organic vapors with later analysis by gas chromatography. Clip unit to your clothing, wear for the duration of the sampling period, and send it to 3M or any other certified gas chromatography lab to determine exposure.
All 3M Air Monitors are simple and effective devices that collect contaminants through the scientific principle of diffusion. They meet or exceed OSHA accuracy requirements of ±25% at 95% confidence level for many work contaminants.
Choose from standard badges and badges with back-up sections. The Organic Vapor Passive Monitoring Badges with Back-Up Section uses primary and back-up sorbent pads for sampling demanding environments. Especially suited for monitoring compounds like acrylonitrile, butadiene and methylene chloride for which activated carbon shows limited capacity.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Analysis Included Gases Measured Description
Availability Pricing
3M Personal Air Monitor, Organic Vapor; 10/Cs WW-60060-98 Mfr # 3M-3500
COMPARE
No Organic Vapor Personal Air Monitor, Organic Vapor; 10/Cs
Call For Price
3M Personal Air Monitor, Organic Vapors; 50/Cs WW-60060-99 Mfr # 3M-3500B
COMPARE
No Organic Vapor Personal Air Monitor, Organic Vapors; 50/Cs
Call For Price
3M Personal Air Monitor, Organic Vapors with Backup; 5/Cs WW-60061-00 Mfr # 3M-3520
COMPARE
No Organic Vapor with Backup Personal Air Monitor, Organic Vapors with Backup; 5/Cs
Call For Price
3M Personal Air Monitor, Organic Vapors with Backup; 50/Cs WW-60061-01 Mfr # 3M-3520B
COMPARE
No Organic Vapor with Backup Personal Air Monitor, Organic Vapors with Backup; 50/Cs