15-Watt Bench Lamps
Ideal for nondestructive testing or contamination control
UV lamps are available in 254 or 365 nm. The 365 nm models have self-filtering, low pressure "black light" tubes which stay cool for a longer period of time than conventional UV tubes making them ideal for non-destructive testing. Highly polished inner aluminum deflector provides maximum UV reflectance.
The 15-watt models are designed for flooding an area of 15 ft² at 20"; the 40-watt models flood an area of 300 ft² at 12 ft. All lamps include two tubes, brackets for wall or ceiling mounting, and a 7-ft, three-wire cord (115 VAC model comes with three-prong plug). Order exposure stand for 15-watt lamps below the table to provide a platform for UV testing.
128,000.00 - ₹134,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Wavelength
Availability Pricing
UV bench lamp; 15 watts, 254 nm, 115 VAC/60 Hz WW-97605-00 Mfr # 95004205
COMPARE
60 115 254 nm
₹128,000.00INR / Each
Analytik Jena UV bench lamp; 15 watts, 254 nm, 230 VAC/50 Hz WW-97605-05 Mfr # 95-0042-09
COMPARE
50 230 254 nm
₹128,000.00INR / Each
Analytik Jena UV bench lamp; 15 watts, 365 nm, 115 VAC/60 Hz WW-97605-10 Mfr # 95004206
COMPARE
60 115 365 nm
₹133,000.00INR / Each
Analytik Jena UV bench lamp; 15 watts, 365 nm, 230 VAC/50 Hz WW-97605-15 Mfr # 95-0042-11
COMPARE
50 230 365 nm
₹134,000.00INR / Each