ZenPure Venting filter disc, 25mm, 50/pk (Female luer/Male luer)
Representative image only.
ZenPure Mfr # D25CF020LFLS-25 – Item # WW-49700-60
Manufactured under strict cleanroom conditions and totally biosafe
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPTFE
  • Pore Size (micron)0.2
  • Filter Size25
  • Housing MaterialPolypropylene
  • ConnectionsFemale luer by Male luer
  • Effective Filtration Area (cm²)4.6
  • Surface Area (m²)0.00046
  • DescriptionVenting filter disc, 25mm, 50/pk (Female luer/Male luer)
More About this Item
Filters feature a polypropylene housing and a 0.2 µm PTFE membrane allowing easy scale-up within the range based on vessel capacity and flow rate required. These filters are ideal for venting and isolation applications to protect vessel contents from external contaminants in biotechnology, pharmaceutical, and food and beverage industries. Filters are also used to protect the environment from contaminants within the vessel.