ZenPure Ground water filter capsule, 5.0um (1/4" to 3/8" stepped hosebarb)
Representative image only.
ZenPure Mfr # SKLP050H2HS2HS – Item # WW-49705-96
Filter groundwater samples for dissolved metal analysis
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPolypropylene
  • Pore Size (micron)5
  • Housing MaterialPolypropylene
  • Connections1/4\" to 3/8\" hose barb
  • Effective Filtration Area (cm²)720
  • Surface Area (m²)0.072
  • DescriptionGround water filter capsule, 5.0um (1/4\" to 3/8\" stepped hosebarb)
More About this Item
Groundwater filter capsules are designed to make the process of collecting groundwater samples for analysis quick, easy, and convenient. Capsules contain no glues, adhesives, or other surfactants. All housing components are thermally fused. Capsules provide at least 720 cm2 of effective filtration area. Connections are 1/4 to 3/8" (6 to 9 mm) stepped barb.