YSI ODO/T Optical Dissolved Oxygen Probe and Cable Assembly, 30 m Cable
Representative image only.
YSI Mfr # 627200-30 – Item # WW-59353-75
Get lab-quality results at the source
  • Advanced high-accuracy optical dissolved oxygen probe requires little maintenance Smart probes store calibration information for up to one year saving you time and money
  • Includes 30-m cable
  • 2 year warranty
PRODUCT OPTIONS
₹226,000.00INR / Each
Include Calibration Solution!
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsWater quality, waste water, natural resources, oxygen analysis
  • Min Temperature (° C)-5
  • Max Temperature (° C)70
  • Temperature CompensationYes
  • GuardStainless steel
  • ConnectorWaterproof
  • Cable Length (m)30
  • DescriptionOptical Dissolved Oxygen Probe and Cable Assembly, 30 m Cable
Two year warranty
Includes
Probes with cables 13-ft (4-m) to 100-ft (30-m) include cable management kit.
Key Features
  • Advanced high-accuracy optical dissolved oxygen probe requires little maintenance Smart probes store calibration information for up to one year saving you time and money
  • Includes 30-m cable
  • 2 year warranty
More About this Item
Interchangeable digital "smart" probes store calibration data for a year, saving you time and calibration solution costs—no need to recalibrate if connected to different meters. Convenient cable management kits are included with probes featuring cable lengths from 13-ft (4-m) to 100-ft (30-m)