YSI 910 COD Chemical Oxygen Demand (COD ) Colorimeter
Representative image only.
YSI Mfr # 250910Y – Item # WW-99550-10
US EPA-approved COD test method
  • Premeasured COD reagents cover three test ranges
  • 430 nm single-wavelength colorimeter is specific to COD testing
  • Digest up to 24 samples simultaneously with thermal reactor
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • TestChemical oxygen demand (COD)
  • DescriptionChemical Oxygen Demand (COD ) Colorimeter
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • Premeasured COD reagents cover three test ranges
  • 430 nm single-wavelength colorimeter is specific to COD testing
  • Digest up to 24 samples simultaneously with thermal reactor
  • 2 year warranty
More About this Item
Testing requires the purchase of all three components: a COD colorimeter, a thermal reactor, and COD test reagents.
YSI 910 COD Colorimeter is set up to directly read the values for the three ranges of COD testing. Meter features IP68-rated waterproof housing, large backlit display, real-time clock, and 16-data set automatic data storage. User calibration possible.