This item is now discontinued. We recommend the following as suggested replacements: WW-94190-08
YSI 500 Electronic Piston Burette; 100-240 VAC, 50/60 Hz
Representative image only.
YSI Mfr # 285220220 – Item # WW-24906-02
Ideal for manual titrations, accurate dosing of small and large volumes and the preparation of solutions
  • Highly visable, full color display can be viewed from a distance
  • Reagent data stored in intelligent exchangeable modules
  • Connection flexibility for printer and/or an analytical balances
  • Complete remote control via RS232 or USB interface
  • 2 year warranty
Often bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Display TypeLCD, 3.5\" H
  • Display Resolution0.001
  • pH Repeatability± 0.15% of span
  • pH Accuracy± 0.15% of span
  • Capacity (mL)20
  • Dispensing Accuracy±0.1 to 0.3% total volume
  • OutputUSB, RS-232
  • Data LoggingYes
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • DescriptionElectronic Piston Burette; 100-240 VAC, 50/60 Hz
  • Warranty2 Years
Two year warrantyISO9001 Certified Supplier
Key Features
  • Highly visable, full color display can be viewed from a distance
  • Reagent data stored in intelligent exchangeable modules
  • Connection flexibility for printer and/or an analytical balances
  • Complete remote control via RS232 or USB interface
  • 2 year warranty
More About this Item
The motor-driven piston burette has intelligent exchangable dosing modules to ensure high precision and method flexibility. System includes preprogrammed standard methods and fifteen user-defined methods. Maximum dosing speed is 100 mL/min.
The intelligent exchangeable modules for with 5, 10, 20, and 50 mL volume store reagent and unit data on an integrated RFID chip for easy changes of titrants/solutions. Burette size (mL), titrant name, titrant concentration, expiration date and more are automatically uploaded with the change of modules.
Flexible configuration with three USB and two RS232 ports for connections with magnetic stirrer, manual controller, printer, keyboard, data storage device, balance, computer, and USB expansion hub for up to 16 devices. Automatic weight recording when balance is connected.