YSI 108132Y IDS Adapter for BNC Electrodes
Representative image only.
YSI Mfr # 108132Y – Item # WW-05510-56
Digital sensors for Plug-and-play capabilities
  • Quality IDS probes with 3-m cable
  • 1 year warranty
PRODUCT OPTIONS
₹51,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Connection Adapters
  • DescriptionIDS Adapter for BNC Electrodes
  • Warranty1 year
1 Year Warranty
Key Features
  • Quality IDS probes with 3-m cable
  • 1 year warranty
More About this Item
Intelligent Design Sensors (IDS) digitally store their serial number, unique ID, and calibration data. The sensors can be moved from one meter to another and transfer their calibration data. Just plug a sensor into a YSI Multilab meter to change the parameter to be measured.
Adapter allows arandard pH and ORP electrodes with a BNC connector to be used with the MultiLab two and three channel meter.