WTW Spectrophotometer for Routine and Spectral Analysis in the VIS Range From 320-1100 nm
Representative image only.
WTW Mfr # 250203 – Item # WW-15961-68
Ideal for wastewater influent and effluent testing, industrial process water testing, and environmental monitoring
  • PhotoLab® 7100 VIS Spectrophotometer covering the range from 320 to 1100 nm. Universal instrument suitable for all routine measurements with test kits, in service labs, industry and R&D, wherever the UV range is t not needed.
  • PhotoLab® 7600 UV-VIS Spectrophotometer covering the range from 190 to 1100 nm.
  • It is Ideal for additional self-control with optical reagent-free methods (OptRF) and UV-VIS procedures for many applications.
  • 1 year warranty
PRODUCT OPTIONS
₹643,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSpectrophotometer for Routine and Spectral Analysis in the VIS Range From 320-1100 nm
  • Warranty1 year
1 Year Warranty
Key Features
  • PhotoLab® 7100 VIS Spectrophotometer covering the range from 320 to 1100 nm. Universal instrument suitable for all routine measurements with test kits, in service labs, industry and R&D, wherever the UV range is t not needed.
  • PhotoLab® 7600 UV-VIS Spectrophotometer covering the range from 190 to 1100 nm.
  • It is Ideal for additional self-control with optical reagent-free methods (OptRF) and UV-VIS procedures for many applications.