WTW 285138192 4,7,10 Buffer Ampoules, 2 x 30 Ampoules Ea
Representative image only.
WTW Mfr # 285138192 – Item # WW-59351-42
Up to 5 years shelf life
  • Convenient ready-to-use, sealed glass ampoules with presterilized buffers
  • Certified accuracy traceable to NIST/DIN standards
  • 2 year warranty
Include Buffers Today!
Oakton® pH Buffer Solution Packs
4,300.00 - ₹44,700.00INR / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Buffer Kit Values4, 7, and 10
  • Volume (mL)25
  • Description4,7,10 Buffer Ampoules, 2 x 30 Ampoules Ea
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • Convenient ready-to-use, sealed glass ampoules with presterilized buffers
  • Certified accuracy traceable to NIST/DIN standards
  • 2 year warranty
More About this Item
Highly precise standards with +/-0.01 pH accuracy. Presterilized and free of preservatives, these single-use buffers offer a 5-year shelf life. Each sealed glass ampoule contains 25 mL of solution.