Worthington R018-8C00 Roller base for Dewars with 15-5/8" dia base
Representative image only.
Worthington Mfr # R018-8C00 – Item # WW-03773-65
  • 2 year warranty
PRODUCT OPTIONS
₹53,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRoller base for Dewars with 15-5/8\" dia base
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Easily move storage Dewars and Dewars with canister storage systems. Sturdy cast aluminum frame has five wheels with ball-bearing swivel casters. Use model 03773-65 for Dewars with 15.6" diameter base; use 03773-71 for Dewars with 18.8" diameter base. Shpg wt 8.0 lb (3.7 kg). Note: Roller base does not include tipping stand.