Winland Electronics EA-WMFS Enviroalert Wireless Multifunction Sensor for Use with 10450-74
Representative image only.
Winland Electronics Mfr # EA-WMFS – Item # WW-10450-77
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionEnviroalert Wireless Multifunction Sensor for Use with 10450-74
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Parts and accessories for your enviornmental monitoring needs.
Multifunction sensor must be used with wireless temperature sensor 10450-74. Additionally, wierless sensors can only be used with 10450-31 (Model EA800).