Wildco 66-C52 Dredging Winch Mount, 2" clamp attaches to sideboards or transoms
Representative image only.
Wildco Mfr # 66-C52 – Item # WW-05475-01
Simple and easy to use - it clamps to the side boards or transom of a small boat
  • Designed for use with 66-C10 winch
  • 3 month warranty
₹63,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDredging Winch Mount, 2\" clamp attaches to sideboards or transoms
  • Warranty3 month
3 Month Warranty
Key Features
  • Designed for use with 66-C10 winch
  • 3 month warranty
More About this Item
Has 4" x 5 1/2" mounting plate for winch 05475-00. Clamps to side boards or transom— 2" clamp opening.