Wildco 66-C10 Dredging Winch, 1000 lb max, 1:1 and 1:5 drives
Representative image only.
Wildco Mfr # 66-C10 – Item # WW-05475-00
Designed for use with any line and instrument weighing up to 1,000 lbs
  • Heavy-duty zinc-plated steel
  • Use requires 66-C52 Utility Winch Mount
  • 3 month warranty
₹99,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyester
  • DescriptionDredging Winch, 1000 lb max, 1:1 and 1:5 drives
  • Warranty3 month
3 Month Warranty
Key Features
  • Heavy-duty zinc-plated steel
  • Use requires 66-C52 Utility Winch Mount
  • 3 month warranty
More About this Item
Handles 1000 lb with 1:1 and 1:5 drives. Drum is 6 1/2" dia; holds 88 ft of 3/16" polyester line or 200 ft of 1/8" aircraft cable. Includes “U” bolts.