DWK Life Sciences (Wheaton) 237070 Alcohol Burner, 100 ml Capacity, 4" H x 3" Dia
Representative image only.
DWK Life Sciences (Wheaton) Mfr # 237070 – Item # WW-48585-84
Ideal for drying slides
  • Can use with isopropyl or denatured ethyl alcohol
  • Low-heat flame
PRODUCT OPTIONS
₹8,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeAlcohol
  • Height (in)4
  • DescriptionAlcohol Burner, 100 ml Capacity, 4\" H x 3\" Dia
Key Features
  • Can use with isopropyl or denatured ethyl alcohol
  • Low-heat flame
More About this Item
Manufactured from soda-lime glass, this burner is designed for use with isopropyl or denatured ethyl alcohol. The low-heat flame is well suited for microscopy purposes.