Wearwell 711.18X3x75BK Smooth Military Approved Switchboard Matting, 3 ft x 75 foot, Black
Representative image only.
Wearwell Mfr # 711.18X3x75BK – Item # WW-55555-44
Protect workers from harmful or deadly electrical shock
  • Military 15562F specifications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialVinyl
  • Width (in)36
  • Width (cm)91.44
  • ColorBlack
  • DescriptionSmooth Military Approved Switchboard Matting, 3 ft x 75 foot, Black
  • Warranty1 year
1 Year Warranty
Key Features
  • Military 15562F specifications
  • 1 year warranty
More About this Item
High-performance, nonconductive vinyl mats insulate workers, protecting them from deadly shocks generated by high-voltage equipment. Mats are tested and proven to have a high dielectric strength; however the maximum recommended voltage exposure is much less.
Smooth Military Switchboard Model 711
Designed to meet strict military standards
Smooth surface offers outstanding ozone and oil resistance and is easy to clean
Passes Federal Flammability standard 501, method 6411
Meets Military Spec 15562-F, Amendment 3, Type II