Wearwell 702.14x75cutbk Corrugated Switchboard Matting, 2' x 75', Black
Representative image only.
Wearwell Mfr # 702.14x2x75BK – Item # WW-55555-14
Protect workers from harmful or deadly electrical shock
  • ASTM D178-01 specifications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialVinyl
  • Thickness (in)0.25
  • Width (in)24
  • Width (cm)60.96
  • ColorBlack
  • DescriptionCorrugated Switchboard Matting, 2' x 75', Black
  • Warranty1 year
1 Year Warranty
Key Features
  • ASTM D178-01 specifications
  • 1 year warranty
More About this Item
High-performance, nonconductive vinyl mats insulate workers, protecting them from deadly shocks generated by high-voltage equipment. Mats are tested and proven to have a high dielectric strength; however the maximum recommended voltage exposure is much less.
Corrugated Switchboard Model 702
Corrugated surface offers additional slip resistance
1/4'' thickness meets Type II, Class 2 specs
3/8'' thickness meets Type II, Class 3 specs
Both thicknesses conforms to ANSI/ASTM D178-01 standards
1/4'' thickness meets Type II, Class 2 specs. Conforms to ANSI/ASTM D178-01 standards.