Wearwell 701.14X4X75BK Corrugated Switchboard Matting, 4' W x 75', Black
Representative image only.
Wearwell Mfr # 701.14X4X75BK – Item # WW-55555-10
Protect workers from harmful or deadly electrical shock
  • ASTM D178-01 specifications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialVinyl
  • Width (in)48
  • Length (ft)75
  • Width (cm)121.92
  • ColorBlack
  • DescriptionCorrugated Switchboard Matting, 4' W x custom size to 75', custom size
  • Warranty1 year
1 Year Warranty
Key Features
  • ASTM D178-01 specifications
  • 1 year warranty
More About this Item
High-performance, nonconductive vinyl mats insulate workers, protecting them from deadly shocks generated by high-voltage equipment. Mats are tested and proven to have a high dielectric strength; however the maximum recommended voltage exposure is much less.
Diamond-Plate Switchboard Model 701
Distinctive diamond-embossed surface is easily cleaned and won't collect contaminants
Conforms to ANSI/ASTM D178-01 Type II, Class 2 Specs