Waterless ice bag kit; 2L beads, two chill packs
Representative image only.
Lab Armor Item # WW-06274-26
Replace water in laboratory water baths and ice in ice buckets.
  • Reduce the risk of contamination, achieve reproducible results and have greater confidence in your work
  • No need to worry about burnout as there is no water to evaporate.
  • Highly recyclable, fewer harmful biocides, and more energy efficient beads are eco-friendly
  • Durable nylon mesh fabric
  • Washable bad that is mildew and rot resistant with a drawstring closure
  • Chill packs are resuable and last for hours
PRODUCT OPTIONS
₹78,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWaterless ice bag kit; 2L beads, two chill packs
Key Features
  • Reduce the risk of contamination, achieve reproducible results and have greater confidence in your work
  • No need to worry about burnout as there is no water to evaporate.
  • Highly recyclable, fewer harmful biocides, and more energy efficient beads are eco-friendly
  • Durable nylon mesh fabric
  • Washable bad that is mildew and rot resistant with a drawstring closure
  • Chill packs are resuable and last for hours
More About this Item
Compatible with waterless ice buckets, items 06274-24 and -28.
Waterless ice bag kit includes a nylon bead bag with 0.5 gallons (2 L) of beads and two chill packs.