Vision USA 6975 Handheld LED Illuminated Magnifier, 2" Dia Viewing Area; 14x
Representative image only.
Vision USA Mfr # 127077 – Item # WW-03889-88
High quality LED illumination
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification14x
  • Viewing Area1.3\" (35 mm) diameter
  • LensSingle
  • Mounting TypeHandheld
  • Light SourceLED
  • BatteryAA Battery (2)
  • DescriptionHandheld LED Illuminated Magnifier, 2\" Dia Viewing Area; 14x
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The large viewing area and brighter light provides greater detail and less eye-strain during inspections.
Handheld models feature a compact, pocket-sized design—ideal for field use. Handheld magnifiers require three AAA batteries (sold separately).