Spex VapLock™ One-Way Stopcock Conector, Female Luer to Male Luer
Representative image only.
Spex Mfr # FCFM-001 – Item # WW-12023-27
Easily restrict or stop the flow of fluids
  • Clear polycarbonate (PC) body
  • Durable polyethylene (PE) handle on top of stopcock
  • Ideal for use in a variety of industries
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolycarbonate body with arm or handle made of polyethylene
  • Number of Ports2
  • Connection Type-Port 1Female Luer
  • Connection Type-Port 2Male Luer
  • DescriptionOne-Way Stopcock Conector, Female Luer to Male Luer
Key Features
  • Clear polycarbonate (PC) body
  • Durable polyethylene (PE) handle on top of stopcock
  • Ideal for use in a variety of industries
More About this Item
The one-way stopcock connector involves a capped threaded female luer to fit into a male slip luer, while the three-way consists of two threaded female luers to fit into a male slip luer.