Vanguard Safety A14 Nitrile Chemo Exam Gloves, Medium; 2000/CS
Representative image only.
Vanguard Safety Mfr # A14C12 – Item # WW-78902-02
Maintain dexterity without sacrificing safety
  • Medical grade and chemotherapy rated
  • Powder-free—made of soft 100% nitrile
  • Latex-free to eliminate adverse reactions
  • Textured surface for better gripping
  • Ambidextrous
  • Gloves are size medium
PRODUCT OPTIONS
₹18,100.00INR / Case of 2000
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)3
  • SizeMedium
  • ColorBlue
  • GripTextured surface for improved grip
  • CuffAmbidextrous Beaded Cuff
  • SterileNo
  • Includes200 gloves/Box, 10 boxes/Case
  • PowderedNo
  • DescriptionNitrile Chemo Exam Gloves, Medium; 2000/Cs
Key Features
  • Medical grade and chemotherapy rated
  • Powder-free—made of soft 100% nitrile
  • Latex-free to eliminate adverse reactions
  • Textured surface for better gripping
  • Ambidextrous
  • Gloves are size medium
More About this Item
These Vanguard Safety A14 nitrile chemo exam gloves are made from powder-free and latex-free nitrile. They feature textured surface for improved grip and maximum ease of use with delicate instruments. All A14 nitrile gloves are ambidextrous and blue in color. Gloves are packed 200/box and 10 boxes per case.