Vaisala HMT360 Duct-Mount Humidity Transmitter without Display

Representative image only.
Vaisala Mfr # HMT3603D12BCD2A4BB5A1B – Item # WW-37510-14
Factory Mutual approved for hazardous indoor applications
  • Meets Class I, II, III, Division 1, Groups A-G and Division 2, Groups A-D, F and G
  • Picture is shown with a display, however product does not come with a display.
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Min RH Range (%)0
  • Max RH Range (%)100
  • RH Accuracy±1% RH; ±2% RH above 90% RH
  • Min Temperature (° F)-40
  • Min Temperature (° C)-40
  • Max Temperature (° F)248
  • Max Temperature (° C)128
  • Temperature Accuracy±0.36°F (±0.2°C)
  • Response Time15 seconds
  • Mounting TypeDuct
  • Output4-20mA
  • Power (VDC)12 to 28
  • Height (in)4 1/2
  • Width (in)6 1/2
  • Depth (in)2 3/8
  • DescriptionDuct-Mount Humidity Transmitter without Display
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 65NIST Traceable Calibration

Key Features

  • Meets Class I, II, III, Division 1, Groups A-G and Division 2, Groups A-D, F and G
  • Picture is shown with a display, however product does not come with a display.
  • 1 year warranty

More About this Item

Measure temperature and humidity in environmentally hazardous areas. NEMA 4 (IP65) housing is rated water- and dust-proof. Wall-mount and duct-mount versions are available with or without an LCD. Two 4 to 20 mA outputs offer analog signals, one for humidity and one for temperature.
What’s included: certificate of NIST traceability. Wall-mount transmitters include a probe measuring 4 5/8”L x 1/2” dia. Duct-mount transmitters include a probe measuring 2 11/16”L x 1/2” dia with 78" (2 m) of cable length, and a duct installation kit.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat