Vaisala HM70D3D1A1DB Thermohygrometer with Hmp77 Remote Probe and Rechargable Batteries
Representative image only.
Vaisala Mfr # HM70D3D1A1DB – Item # WW-37850-18
Dual probe input for multi-parameter measurement
  • Store 2700 data points—download later to your PC via RS-232 interface
  • Display measurements numerically or graphically
  • Three probe types available; tailor the meter for your application
  • Includes
  • MI70Link software with RS232 and analog output signal cables
  • Connection cable to HMD/W60/70 series
  • HMP77 remote small sensor humidity probe with 5 meter cable
  • Weather proof carrying case
  • Rechargable batteries
PRODUCT OPTIONS
Call For Price
₹29,500.00 - ₹37,500.00INR / Each
for an additional
₹29,500.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Data LoggingYes
  • Battery1 Rechargeable NiMH (included)
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionThermohygrometer with Hmp77 Remote Probe and Rechargable Batteries
Includes
MI70Link software with RS232 and analog output signal cables, connection cable, HMP77 remote small sensor humidity probe with 5 meter cable, weather proof carrying case, and 1 rechargeable NiMH battery.
Key Features
  • Store 2700 data points—download later to your PC via RS-232 interface
  • Display measurements numerically or graphically
  • Three probe types available; tailor the meter for your application
  • Includes
  • MI70Link software with RS232 and analog output signal cables
  • Connection cable to HMD/W60/70 series
  • HMP77 remote small sensor humidity probe with 5 meter cable
  • Weather proof carrying case
  • Rechargable batteries
More About this Item
Vaisala HM70 series meter measures temperature and humidity, plus calculates dew point, wet bulb, mixing ratio, absolute humidity, enthalpy, and water vapor pressure. It offers six different languages in display. Log three parameters into meter memory while displaying data in numerical or graphical fashion. View graphic trends of any parameter on the backlit LCD.
Customize your application with three humidity probes (sold separately). The general-purpose probe measures ambient conditions. Use the 12” stainless steel probe for air duct measurements. The extension cable probe includes a humidity sensor at the end of a 5-meter cable for high-temperature and hard-to-reach measurements. Order probes separately.