Vacuum Pump Tubing, stainless steel, NW 16, 3/4"ID X 20"L
Representative image only.
Item # WW-79752-32
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")20
  • Fitting MaterialStainless Steel
  • Connection TypeVacuum Flange NW16
  • Barb Size / Tubing Id (Port 1) (in)3/4
  • Flange Size (Port 1)NW 16
  • Length (ft)1.67
  • DescriptionVacuum Pump Tubing, stainless steel, NW 16, 3/4\"ID X 20\"L
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Tubing butt-welded seam eliminates leaks and prevents contamination.