UVP 97-0016-04(UVX31) Ultraviolet Intensity Radiometer Sensor, 302 nm
Representative image only.
Analytik Jena Mfr # 97-0016-04(UVX31) – Item # WW-97651-24
With interchangeable NIST-traceable sensors
  • 1 year warranty
PRODUCT OPTIONS
₹104,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Wavelength302
  • DescriptionUltraviolet Intensity Radiometer Sensor, 302 nm
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This radiometer is used to monitor the useful life of any UV light source by measuring lamp irradiance. It operates with a 9 V battery for up to 120 hours. Select three intensity ranges: 0 to 20 mW/cm2, 0 to 2000 µW/cm2, and 0 to 200 µW/cm2. Optional 10:1 attenuator extends the reading range up to 200 mW/cm2.
Choose one of the three interchangeable sensors for measuring 254 nm, 302 nm, or 365 nm ultraviolet wavelengths (sold separately). All sensors are calibrated with NIST traceability, and are ±5% accurate. Sensors must be purchased separately. The 3-ft cable length is ideal for measuring UV in remote locations.