Uvex by Honeywell SVP200 Safety Glasses, Clear Frame, Clear Hardcoat Lens; 10/Bx; 10/Bx
Representative image only.
Uvex by Honeywell Mfr # SVP200 – Item # WW-86497-71
Premium look, safety, and comfort at an economic price
  • Choose the frame color to suit your tastes Cushioned nose and ear grips for superior comfort and stay Clear polycarbonate lenses are ANSI Z.81.1+2015 certified Select from Hardcoat/anti-scratch or Uvextra Anti-Fog lens coatings 200-series features thin frame vs 400-series thicker frame
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorClear
  • Lens ColorClear
  • Lens CoatingHardcoat anti-scratch
  • Lens TypePolycarbonate
  • ComplianceANSI Z.81.1+2015; CSA 2015
  • DescriptionSafety Glasses, Clear Frame, Clear Hardcoat Lens; 10/Bx
Key Features
  • Choose the frame color to suit your tastes Cushioned nose and ear grips for superior comfort and stay Clear polycarbonate lenses are ANSI Z.81.1+2015 certified Select from Hardcoat/anti-scratch or Uvextra Anti-Fog lens coatings 200-series features thin frame vs 400-series thicker frame
More About this Item
200-series feature thinner frame and also meet CSA 2015 certifications.