US Industrial 8100R Analog Gas Detector/Alarm, Combustible Gas, Wall Mt w/remote sensor; 0-100%LEL

Representative image only.
US Industrial Mfr # 8100R – Item # WW-10110-22
Don’t risk workers’ safety
  • Continuous 24 hour monitoring Compact, durable units Explosion proof housing for sensor Audio/visual alarms Malfunction LED and test/checking switches Infrequent maintenance and calibration required Remote sensor reliable to 600 feet from controller Relay contacts and recorder terminals
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Gases MeasuredCombustibles
  • Min Combustible Range (% LEL)0
  • Max Combustible Range (% LEL)100
  • AlarmAlarm light (red), fail light (yellow), and audible alarm
  • Display TypeAnalog
  • Power (VDC)24
  • Power (VAC)115
  • Height (in)8
  • Width (in)5
  • Depth (in)4
  • Height (cm)20.3
  • Width (cm)12.7
  • Depth (cm)10.2
  • DescriptionAnalog Gas Detector/Alarm, Combustible Gas, Wall Mt w/remote sensor; 0-100%LEL
  • Warranty1 year
1 Year Warranty

Key Features

  • Continuous 24 hour monitoring Compact, durable units Explosion proof housing for sensor Audio/visual alarms Malfunction LED and test/checking switches Infrequent maintenance and calibration required Remote sensor reliable to 600 feet from controller Relay contacts and recorder terminals
  • 1 year warranty

More About this Item

The wall mount unit is for use in an enclosed area. Remote sensor provides open space monitoring ability. At hazardous locations, explosion proof sensors and their junction box housing meet NEC requirements for Class 1, Division 1, Group A-B-C-D. The unit is designed for those who are concerned about explosive and toxic gases over a long period of time mostly within an enclosed area and/or work environment.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat