Idex Flangeless Male Nut, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Representative image only.
Idex Mfr # P-330X – Item # WW-02007-66
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Natural PEEK
  • 10/PK
  • 1 month warranty
PRODUCT OPTIONS
₹6,600.00INR / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)1/4-28
  • Thread size UNF(M) (port 2) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 2) (in)1/8
  • ColorNatural
  • DescriptionFlangeless Male Nut, 1/4-28 UNF for 1/8\" OD Tubing, Peek, 10/Pk
1 Month Warranty
Key Features
  • Removable and reusable—saving you money!
  • Natural PEEK
  • 10/PK
  • 1 month warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
Flangeless Fittings are easy to assemble. Slide the ferrule over the tubing and fingertighten
the nut. Excellent chemical resistance. 1/8" fittings are rated to 500 psi; 1⁄16" fittings
are rated to 1000 psi. All fittings feature 1⁄4-28 threads.