Two-position cap; size 17mm; color, natural; material, PE; 1000/pack
Representative image only.
Item # WW-63600-65
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyethylene(PE)
  • Cap MaterialPolyethylene (PE)
  • Cap / Stopper Size17 mm
  • Cap ColorNatural
  • DescriptionTwo-position cap; size 17mm; color, natural; material, PE; 1000/pack
More About this Item
Two-position caps for culture tubes; PE, non-sterile. Two-position caps can be vented or fully closed.