Tunair SS-7001 Sample Shake Flask Kit, 300 mL and 2.5 L
Representative image only.
Tunair Mfr # SS-7001 – Item # WW-01835-07
Increase culture yields with optimum growth conditions for aerobic organisms
  • Baffle and turbo vane closure designs increase oxygenation compared to standard Erlenmeyer flasks
  • Choose between unbaffled, half-baffled and full-baffled flasks based on your application
  • Cap with removable filter lining protects contents of flask from airborne particulates and eliminates need to flame neck
  • All components are autoclavable
  • Suitable for culturing aerobic microbes, mammalian, and plant cells
₹62,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Cap MaterialPolypropylene (PP)
  • DescriptionSample Shake Flask Kit, 300 mL and 2.5 L
Key Features
  • Baffle and turbo vane closure designs increase oxygenation compared to standard Erlenmeyer flasks
  • Choose between unbaffled, half-baffled and full-baffled flasks based on your application
  • Cap with removable filter lining protects contents of flask from airborne particulates and eliminates need to flame neck
  • All components are autoclavable
  • Suitable for culturing aerobic microbes, mammalian, and plant cells
More About this Item
These shake flasks work with any standard orbital shaker having adjustable speeds from 150 to 400 rpm. The unbaffled models behave similarly to Erlenmeyer flasks under these conditions. The half-baffled flasks create the liquid motion of a vortex, and provide a moderate increase in dissolved oxygen. For the greatest increase in dissolved oxygen, the full-baffled flasks create a motion similar to that of an overhead stirrer with an axial impeller.
Cleanup is easy — just soak in a mild dish soap to remove dirt, and air dry. All pieces are autoclavable, including the filter linings. Filter linings are also replaceable, with silicone membrane and Dri-Gauze materials available.