TSI 794 VelociCalc Air Temperature Probe
Representative image only.
TSI Mfr # 794 – Item # WW-10372-41
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-40
  • Max Temperature (° F)1600
  • Min Temperature (° C)-40
  • Max Temperature (° C)870
  • Temperature Accuracy±0.1% of reading +2°F (±0.056% of reading +1.1°C)
  • Temperature Resolution0.1°F (0.1°C)
  • DescriptionVelociCalc Air Temperature Probe
  • Warranty2 year
Two year warrantyNIST Traceable Calibration
Key Features
  • 2 year warranty
More About this Item
Keep your TSI ventilation meter working dependably with these parts and accessories.