Trier Solid Sampler, 17 mL capacity, 7"L
Representative image only.
Item # WW-33687-36
Sharp enough to cut through a bag or box
  • Ideal for sampling solids such as cheese, grease, or margarine
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (cm)17.7
  • Diameter (cm)1.5
  • DescriptionTrier Solid Sampler, 17 mL capacity, 7\"L
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for sampling solids such as cheese, grease, or margarine
  • 1 year warranty
More About this Item
To extract sample, insert the sharp end of the sampler into the solid material. Once inserted, twist the handle one-half turn. This will cut out a conical plug of material. The produced hole on the side of the material may be closed by applying adhesive tape or wax. Sampler is made of stainless steel and can be sterilized. Conical sampler, including handle, measures 177 mm L x 15 mm dia (11 mm at the tapered end).