Trajan SGE 033012 Replacement Kit, for 0.5 uL Plunger-in-Needle Syringe, 50 mm L, 0.47 mm OD, CT; 1/Ea
Representative image only.
Trajan Mfr # 033012 – Item # WW-16002-99
Choose wisely to save your operational cost—no need to buy new syringes instead
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTrajan SGE Replacement Kit, for 0.5 uL Plunger-in-Needle Syringe, 50 mm L, 0.47 mm OD, CT; 1/Ea
  • Warranty6 month
6 Month Warranty
Key Features
  • 6 month warranty
More About this Item
Trajan SGE Syringe Repair Kits are available for 0.5 to 5.0 µL plunger-in-needle syringes. The kits are supplied with both a plunger and a needle, both must be replaced together. The replacement kits are listed below to determine the appropriate replacement plunger-in-needle kit.
Needle Selection:
• Needle length refers to the exposed length of the needle when fitted to syringe
• Needle gauge is nominal, please refer to OD and ID
• Choose a different replacement needle by selecting the syringe volume, needle length, OD and tip style
Plunger Selection:
• All SGE gas-tight syringes have replaceable plungers. The replacement plungers are listed below to determine the appropriate replacement plunger.
• The standard plunger in MicroVolume syringes is not replaceable; each stainless steel plunger is individually fitted to each barrel.