Trajan SGE 006000 Microvolume Syringe with Fixed Bevel-Tip Needle and Plunger, 250 uL, 50 mm L, 25 G; 1/Ea
Representative image only.
Trajan Mfr # 006000 – Item # WW-16001-07
Designed for a vast variety of applications with outstanding performance and superior quality
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTrajan SGE Microvolume Syringe with Fixed BT Needle and Plunger, 250 uL, 50 mm L, 25 G; 1/Ea
  • Warranty6 month
6 Month Warranty
Key Features
  • 6 month warranty
More About this Item
Trajan SGE Syringes have consistently been the choice of global GC & LC manufacturers because of outstanding performance and superior quality in a vast variety of applications. The unique Plunger tip designed ensures a gas-tight seal providing the lowest syringe carryover and outstanding reproducibility.
Available range includes following variants:
GC Autosampler Syringes:
• SGE range of GC Autosampler syringes are designed and tested to meet critical autosampler specifications
• Wide range of fixed and removable needle options
• PTFE tipped replaceable plunge for gas and headspace samples
Gas-Tight Syringes:
• Constructed of borosilicate glass and interchangeable PTFE plunger tips
• Maximum inertness for liquid and gas samples
• Capacity varies from 10 µL to 10 mL
• High accuracy of dispensed volume
Jumbo Syringes:
• Designed for holding and dispensing large volumes of gas
• Heavy-duty acrylic barrels
• Easy access to sample for the addition of standards or removal of subsample via secondary port
• Plunger stem can be unscrewed for ease of transportation and storage
• Available in 500 mL, 1 L and 2 L volumes