Traceable 2 Year Premium TraceableLive® Subscription Activation Code
Representative image only.
Traceable Item # WW-63003-39
Simplified cloud monitoring
  • TraceableLIVE-capable products securely monitor your critical environments and notify your smartphone, tablet, or PC if anything goes wrong
  • Notifications for peace of mind - everyone on your team can receive e-mail, text*, voice*, and push notifications on the environments you monitor
  • Remotely view data logging history, effortlessly generate reports, and control alarm parameters
  • Traceable® products come calibrated to NIST standards, helping you meet quality and regulatory requirements
  • Digital data reports sent with ease whenever you need them
  • Customize your report with location, device, date range, and report format
  • 2 year premium subscription
PRODUCT OPTIONS
₹36,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Operating PlatformTraceableLIVE®
  • Description2 Year Premium TraceableLive® Subscription Activation Code
Includes
Card with unique Activation Code and QR code. Note: This Activation Code is good for one (1) TraceableLIVE device only. Every TraceableLIVE device will require one (1) Activation Code to apply Standard Features.
Key Features
  • TraceableLIVE-capable products securely monitor your critical environments and notify your smartphone, tablet, or PC if anything goes wrong
  • Notifications for peace of mind - everyone on your team can receive e-mail, text*, voice*, and push notifications on the environments you monitor
  • Remotely view data logging history, effortlessly generate reports, and control alarm parameters
  • Traceable® products come calibrated to NIST standards, helping you meet quality and regulatory requirements
  • Digital data reports sent with ease whenever you need them
  • Customize your report with location, device, date range, and report format
  • 2 year premium subscription
More About this Item
Purchase a subscription code for 1 or 2 years of TraceableLIVE® (Code applies upgraded features to 1 TraceableLIVE device). Subscriptions are available in standard or premium. Scan the QR code to begin setup or go to traceablelive.com/activate and enter the unique 11-digit number on the card.
A standard subscription activation code includes the following features: E-mail and Push Alarms, Unlimited Data Storage, Unlimited Team Members, and Calibration Due notification.
A premium subscription includes the following features: E-mail and Push Alarms, Free SMS Text Notifications, Unlimited Data Storage, Unlimited Team Members, Calibration Due notification, and 21 CFR Part 11 Compliance.
TraceableLIVE® Fully Compliant Solution meets 21 CFR Part 11 North American compliance and Annex 11 for Europe. All compatible devices include ISO 17025 accredited calibration. If any of your TraceableLIVE® devices detect an out of range condition, lose connection to your network, begin to run low on battery, or if your device is due for calibration TraceableLIVE® will immediately send you alerts based on your settings.
*Text and Voice alerts available with Premium Subscription Activation Codes Only