Traceable Calibrated Thermocouple Thermometer, Type K/J
Representative image only.
Traceable Item # WW-20250-01
Perfect for the lab to production floor
  • Includes A2LA accreditied NIST-Traceable Calibration Certificate
  • Reliable measurements, guaranteed
  • Relative time stamp provides a time reference for major events
  • Electronic off set function allows compensation of thermocouple probe errors
  • 2 year warranty
PRODUCT OPTIONS
₹14,800.00INR / Each
Time to recalibrate?
for an additional
₹19,800.00INR / Each
for an additional
₹27,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels1
  • Types Of Probes AcceptedType J, Type K
  • Min Temperature (° F)-346
  • Max Temperature (° F)2501
  • Min Temperature (° C)-210
  • Max Temperature (° C)1372
  • Accuracy±0.5% of reading +1.8°F; ±0.5% of reading +3.6°F below -148°F [±0.5% of reading +1.0°C; ±0.5% of reading +2.0°C below -100°C]
  • Resolution0.1°; 1° above 1000°
  • Data LoggingNo
  • Low Battery IndicatorYes
  • Display TypeBacklit LCD
  • Height (in)6 3/8
  • No. of Probe(s) Included1
  • Battery3 AAA (included)
  • DescriptionCalibrated Thermocouple Thermometer, Type K/J
  • Warranty2 Years
Two year warrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Includes A2LA accreditied NIST-Traceable Calibration Certificate
  • Reliable measurements, guaranteed
  • Relative time stamp provides a time reference for major events
  • Electronic off set function allows compensation of thermocouple probe errors
  • 2 year warranty
More About this Item
Handheld thermocouple thermometers provide accurate and reliable temperature measurement in an economic and compact design. Features include Min/Max/Avg, Hold, °F/°C or Kelvin (K) selectable units, relative time stamp on Min/Max/Avg, electronic off set function and auto power-off to save on battery power. Meters accept both type K or J thermocouple probes with a miniconnector.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA.  It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Model 20250-01 has a single type K or J input and unique side-button controls for easy one-handed operation. What's included: Type K bead wire temperature probe, three AAA batteries, and NIST-traceable calibration report with data supplied by InnoCal.
Guaranteed to have at least one year of calibration remaining on certificate.