Traceable Replacement Platinum RTD, Stainless-Steel Probe for 37804-07
Representative image only.
Traceable Item # WW-37804-09
Optimize performance of your thermometer
  • Replacement probe for your Traceable Scientific RTD thermometer
  • 10' PTFE cable
  • Traceable® model number 6423
  • 1 year warranty
PRODUCT OPTIONS
₹39,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-58
  • Max Temperature (° F)302
  • Min Temperature (° C)-50
  • Max Temperature (° C)150
  • Probe Diameter (in)0.16
  • DescriptionCalibrated Replacement Platinum RTD, Stainless-Steel Probe for 37804-07
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Replacement probe for your Traceable Scientific RTD thermometer
  • 10' PTFE cable
  • Traceable® model number 6423
  • 1 year warranty
More About this Item
Replace your probe or keep a convenient spare.
For improved performance, probes should be calibrated as a system with the meter.