Traceable Printing Thermocouple Thermometer with Calibration
Representative image only.
Traceable Item # WW-37803-89
Printing thermometer provides permanent record, prints time and date
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Range: –328 to 2431°F and –200 to 1333°C
  • Resolution: 0.1°, accuracy: ±1°C from –50 to 982°C, otherwise ±1.5 °C
  • Prints the time and date for MAX only, MIN only, average only, and in circularly mode T1, T2 and differential, printing programs in intervals of 3 secs to 23h 59m 59s
  • Memory, alarm (sounds when temperature rises above or falls below set points)
  • Dual probe inputs allow displaying the temperature of probe 1, probe 2, or the difference (delta) of probe 1 and 2 (accepts Type- J, K, T, or E probe)
  • Size: 7.5 x 3 x 1.5"; weight: 10 oz
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Connector TypeMini ANSI
  • DescriptionTraceable® Printing Thermocouple Thermometer with Calibration
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Range: –328 to 2431°F and –200 to 1333°C
  • Resolution: 0.1°, accuracy: ±1°C from –50 to 982°C, otherwise ±1.5 °C
  • Prints the time and date for MAX only, MIN only, average only, and in circularly mode T1, T2 and differential, printing programs in intervals of 3 secs to 23h 59m 59s
  • Memory, alarm (sounds when temperature rises above or falls below set points)
  • Dual probe inputs allow displaying the temperature of probe 1, probe 2, or the difference (delta) of probe 1 and 2 (accepts Type- J, K, T, or E probe)
  • Size: 7.5 x 3 x 1.5"; weight: 10 oz
  • 1 year warranty
More About this Item
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.