Traceable Calibrated Thermocouple Thermometer with Data Output
Representative image only.
Traceable Item # 3780391
Quickly obtain measurement readings 2.5 times per second
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Temperature range is –50 to 1999°F and –50 to 1300°C
  • Resolution is 0.1° from –50.0 to 99.9° and 1° elsewhere, accuracy is ±0.3% plus 1°C
  • Maximum only/HOLD
  • Size: 2.75 x 5.25 x 1.25"; weight: 5.5 oz
  • Traceable® model number 4134
  • 1 year warranty
₹20,700.00INR / Each
Time to recalibrate?
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Temperature range is –50 to 1999°F and –50 to 1300°C
  • Resolution is 0.1° from –50.0 to 99.9° and 1° elsewhere, accuracy is ±0.3% plus 1°C
  • Maximum only/HOLD
  • Size: 2.75 x 5.25 x 1.25"; weight: 5.5 oz
  • Traceable® model number 4134
  • 1 year warranty
More About this Item
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Guaranteed to have at least one year of calibration remaining on certificate.
Download your certificate of traceability at certificates.traceablelive.com