Traceable Calibrated Four-Channel Alarm Timer
Representative image only.
Traceable Item # WW-90225-35
Large display allows viewing from across the lab
  • Memory returns display to previously programmed countdown time with touch of a button
  • Extra-loud alarms sound for 1 minute
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Traceable® model number 5004
  • 1 year warranty
₹4,700.00INR / Each
Upgrade version available...
Traceable Four-Channel Timer
₹4,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Timing Capacity100 hours
  • Number of Channels4
  • Resolution1 second
  • Accuracy±0l.5 sec/day
  • DisplayLCD
  • Battery Type1 1.5V Lithium (included)
  • DescriptionCalibrated Four-Channel Alarm Timer
  • Warranty1 year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Memory returns display to previously programmed countdown time with touch of a button
  • Extra-loud alarms sound for 1 minute
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Traceable® model number 5004
  • 1 year warranty
More About this Item
This timer allows you to count up to or down from 100 hours with 1-second resolution. This timer features a 3/4" high display and multiple channels that can be used simultaneously. Each channel has a distinctive alarm that sounds for 1 minute, pressing any button will turn off the alarm. Timer memory recalls previous time set. Features time-of-day in 12- or 24-hour formats.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Guaranteed to have at least one year of calibration remaining on certificate.