Traceable 6602 Calibrated Replacement Bottle Probe with Calibration
Representative image only.
Traceable Item # WW-99460-02
Save time and money by simply replacing probes that are damaged or due for calibration
  • No need to send in your thermometer for re-calibration—simply replace the recalibrated probe
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • Suitable for use with thermometer 99460-01
  • Traceable® model number 6602
  • 1 year warranty
PRODUCT OPTIONS
₹11,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlycol-filled bottle probe
  • Compatible With99460-01
  • Length (in)120
  • DescriptionReplacement Bottle Probe with Calibration
  • Warranty1 Year
1 Year WarrantyMade in the U.S.A.NIST Traceable Calibration
Key Features
  • No need to send in your thermometer for re-calibration—simply replace the recalibrated probe
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • Suitable for use with thermometer 99460-01
  • Traceable® model number 6602
  • 1 year warranty
More About this Item
These replacement temperature probes are compatible with the Traceable® Data Logging Ethernet Thermometers.
Traceable® products are calibrated to NIST-traceable standards and ready to use as soon as you receive it. Each product goes through an intense vetting process before it can carry the Traceable name, and it includes NIST-traceable certification. This means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving you both time and money by not having to calibrate separately.
NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
Nontoxic glycol-filled bottle probe protecting them from rapid temperature changes when refrigerator door is opened.
Probe is suitable for use with the Traceable 6600 Data Logging Ethernet Thermometer