Chem Express X521-25 OV/IAQ Badge with PrePaid Analysis PK/5
Representative image only.
Chem Express Mfr # X521-25 – Item # WW-86378-13
Analysis performed in an AIHA-accredited lab
  • Save money and time with prepaid analysis
  • Lightweight and easy to use
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gases MeasuredOrganic vapors
  • DescriptionOV/IAQ Badge with PrePaid Analysis PK/5
  • Warranty1 year
1 Year Warranty
Key Features
  • Save money and time with prepaid analysis
  • Lightweight and easy to use
  • 1 year warranty
More About this Item
TraceAir® OSP / IAQ Monitor Badge is a high sampling rate monitor used to address Indoor Air Quality concerns. Use when organic vapors are suspected to be the cause of IAQ concern. The Organic Solvent Panel (OSP) badge includes analysis by GC/FID for 25 common organic vapors.
Personal monitoring badges are the most cost effective and efficient ways to monitor chemicals in your environment. Badges are easy to use and require minimal training and experience. They are small and light weight and have minimal effect on worker productivity. Badges are diffusive devices that require no additional equipment such as sampling pumps, calibration materials, or chargers. These long term badges have a low sampling rate preventing the exceedance of their capacity. Analysis on all badges is pre